Learn More
SCAT™ Safety Funnel 'MARCO' with Lid, V2.0
Can be safely closed to prevent danger of ignition.
128.00 € - 341.00 €
Specifications
Diameter (Metric)Top 140 mm
Color Black
View More Specs
Products 15
Product Code Brand Product Type Thread Size (Metric) Includes Price Quantity & Availability  
Product Code Brand Product Type Thread Size (Metric) Includes Price Quantity & Availability  
16852130
View Documents
SCAT™
318962
Safety Funnel with Hinged Lid, Type 2 GL45 Funnel with Lid, Removable Sieve, Earthing Cable
132.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
17155501
View Documents
SCAT™
306980
Safety Funnel with Hinged Lid, Type 1 GL45 Funnel with Lid, Removable Sieve, Earthing Cable
132.00 €
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
16802140
View Documents
SCAT™
318985
Safety Funnel with Hinged Lid S50 Lance (220 mm), Funnel with Lid, Removable Sieve, Earthing Cable
137.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
16892130
View Documents
SCAT™
318983
Safety Funnel with Hinged Lid S51 Lance (220 mm), Funnel with Lid, Removable Sieve, Earthing Cable
137.00 €
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
16842130
View Documents
SCAT™
318961
Safety Funnel with Hinged Lid, Type 1 S55 Funnel with Lid, Removable Sieve, Earthing Cable
136.00 €
Each
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
16882130
View Documents
SCAT™
318981
Safety Funnel with Hinged Lid, Type 2 S55 Lance (220 mm), Funnel with Lid, Removable Sieve, Earthing Cable
137.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
16832130
View Documents
SCAT™
318960
Safety Funnel with Hinged Lid, Type 1 S60/61 Funnel with Lid, Removable Sieve, Earthing Cable
132.00 €
Each
Estimated Shipment: 23-04-2025
to see stock.
Add to basket
16872130
View Documents
SCAT™
318980
Safety Funnel with Hinged Lid, Type 4 S60/61 Lance (220 mm), Funnel with Lid, Removable Sieve, Earthing Cable
137.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
16812130
View Documents
SCAT™
317651
Safety Funnel with Hinged Lid, Type 3 S60/61 Funnel with Lid, Removable Sieve, Earthing Cable, Level Control with 190 mm Floater
341.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
16802130
View Documents
SCAT™
317637
Safety Funnel with Hinged Lid, Type 2 S60/61 Funnel with Lid, Removable Sieve, Earthing Cable, Level Control with 150 mm Floater
341.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
16862130
View Documents
SCAT™
318964
Safety Funnel with Hinged Lid, Type 1 S65 Funnel with Lid, Removable Sieve, Earthing Cable
132.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
16872140
View Documents
SCAT™
318984
Safety Funnel with Hinged Lid S65 Lance (220 mm), Funnel with Lid, Removable Sieve, Earthing Cable
137.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
17863530
View Documents
SCAT™
317639
Safety Funnel with Hinged Lid S70/71 Funnel with Lid, Removable Sieve, Earthing Cable, Level Control with 150 mm Floater
341.00 €
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
17208870
View Documents
SCAT™
318630
Safety Funnel with Hinged Lid, Double Thread R2 in. BSP/G2 (m) and 2 in. Tri-Sure (m) Funnel with Lid, Removable Sieve, Earthing Cable
132.00 €
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17957250
View Documents
SCAT™
318965
Safety Funnel with Hinged Lid Funnel with Lid, Removable Sieve, Earthing Cable
128.00 €
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
Description
Description
  • Modern lid funnel made of high-quality HDPE with very high resistance to acids, alkalis and many other chemicals
  • Integrated gripping edge on funnel lid increases grip without having to touch possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes funnel edge to prevent escape of explosive or health-endangering gases
  • Sturdy hinge guarantees long service life of the closing mechanism
  • Rotating screw cap makes it easier to screw lid funnel on and off the canister
  • Removable sieve for easy cleaning
  • Conductive version of MARCO V2.0 is black, differring in color from the non-conductive version
Specifications
Specifications
140 mm
Black
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
SCAT™ Safety Funnel 'MARCO' with Lid, V2.0