missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Safety Funnel 'MARCO' with Lid, V2.0

Can be safely closed to prevent danger of ignition.

Brand:  SCAT™ 317651

269.36 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 16812130

  • 341.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Modern lid funnel made of high-quality HDPE with very high resistance to acids, alkalis and many other chemicals
  • Integrated gripping edge on funnel lid increases grip without having to touch possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes funnel edge to prevent escape of explosive or health-endangering gases
  • Sturdy hinge guarantees long service life of the closing mechanism
  • Rotating screw cap makes it easier to screw lid funnel on and off the canister
  • Removable sieve for easy cleaning
  • Conductive version of MARCO V2.0 is black, differring in color from the non-conductive version
Specifications

Specifications

140 mm
HDPE, Conductive
Not Autoclavable
Funnel with Lid, Removable Sieve, Earthing Cable, Level Control with 190 mm Floater
Safety Funnel with Hinged Lid, Type 3
300 mL
370 mm
Black
S60/61
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SCAT™ Safety Funnel 'MARCO' with Lid, V2.0 > Safety Funnel V2.0; Thread Size (Metric): S60/61; Includes: Funnel with Lid, Removable Sieve, Earthing Cable, Level Control with 150 mm Floater; Product Type: Safety Funnel with Hinged Lid, Type 3; Material: HDPE, Conductive; Diameter (Metric) Top: 140 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.