Learn More
Thermographic Measurements Ltd Thermax Encapsulated Temperature Indicator
Single use, irreversible threshold indicator reports the highest temperature reached
21.75 € - 25.05 €
Specifications
Certifications/Compliance | ISO 9001 |
---|---|
Disposable | Single-use |
For Use With (Application) | For laboratory research, medical services, instrumentation, electronics industry, automotive, aerospace, steel, metallurgy, plastics industry, food industry applications |
Height (English) | 0.51 in. |
Height (Metric) | 13 mm |
Product Code | Brand | Temperature Range (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Price | Quantity & Availability | ||||||
11770909
|
Thermographic Measurements Ltd
M01ETHE040D01P1 |
40°C |
23.19 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11710919
|
Thermographic Measurements Ltd
M01ETHE049D01P1 |
49°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11720919
|
Thermographic Measurements Ltd
M01ETHE054D01P1 |
54°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11730919
|
Thermographic Measurements Ltd
M01ETHE060D01P1 |
60°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11740919
|
Thermographic Measurements Ltd
M01ETHE065D01P1 |
65°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11750919
|
Thermographic Measurements Ltd
M01ETHE071D01P1 |
71°C |
21.75 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
11760919
|
Thermographic Measurements Ltd
M01ETHE077D01P1 |
77°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11790919
|
Thermographic Measurements Ltd
M01ETHE093D01P1 |
93°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11780919
|
Thermographic Measurements Ltd
M01ETHE088D01P1 |
88°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11720929
|
Thermographic Measurements Ltd
M01ETHE110D01P1 |
110°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11740929
|
Thermographic Measurements Ltd
M01ETHE121D01P1 |
121°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11750929
|
Thermographic Measurements Ltd
M01ETHE127D01P1 |
127°C |
21.75 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11700939
|
Thermographic Measurements Ltd
M01ETHE154D01P1 |
154°C |
21.75 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
11770929
|
Thermographic Measurements Ltd
M01ETH138D01P1 |
138°C |
21.75 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
11710939
|
Thermographic Measurements Ltd
M01ETHE160D01P1 |
160°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11720939
|
Thermographic Measurements Ltd
M01ETHE166D01P1 |
166°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11730939
|
Thermographic Measurements Ltd
M01ETHE171D01P1 |
171°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11740939
|
Thermographic Measurements Ltd
M01ETHE177D01P1 |
177°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11760939
|
Thermographic Measurements Ltd
M01ETHE188D01P1 |
188°C |
25.05 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
11790939
|
Thermographic Measurements Ltd
M01ETHE204D01P1 |
204°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11710949
|
Thermographic Measurements Ltd
M01ETHE216D01P1 |
216°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11720949
|
Thermographic Measurements Ltd
M01ETHE224D01P1 |
224°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11740949
|
Thermographic Measurements Ltd
M01ETHE241D01P1 |
241°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11730949
|
Thermographic Measurements Ltd
M01ETHE232D01P1 |
232°C |
25.05 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
11760949
|
Thermographic Measurements Ltd
M01ETHE254D01P1 |
254°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11750939
|
Thermographic Measurements Ltd
M01ETHE182D01P1 |
182°C |
25.05 €
Pack of 50 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
Description
Specifications
ISO 9001 | |
For laboratory research, medical services, instrumentation, electronics industry, automotive, aerospace, steel, metallurgy, plastics industry, food industry applications | |
13 mm | |
12 mm | |
Wax-melt Technology | |
Irreversible Temperature Indicator | |
Yes | |
Single Temperature |
Single-use | |
0.51 in. | |
40 to 60% | |
2 to 5 seconds | |
9 g | |
0.47 in. | |
Electrical equipment, pumps, braking systems, water pumps, aeronautical equpment, military equipment | |
Dual °C (°F) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermographic Measurements Ltd Thermax Encapsulated Temperature Indicator