missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue, 1% w/v aq. soln.

Brand:  Thermo Scientific Chemicals 042771.AP

50.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11306159

  • 50.75 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Methylene Blue, 1% w/v aqueous solution is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It is also used in the sulfide analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Methylene Blue
1% w/v aqueous solution
MFCD00012111
319.86
23 hPa (17mm Hg) at 20°C
Liquid
122965-43-9, 7732-18-5
C16H18ClN3S
Miscible with ethanol,chloroform,glacial acetic acid and glycerol. Slightly miscible with pyridine. Immiscible with ethyl ether,xylene,oleic acid,ethanol and acetone.
3, 7-Bis(dimethylamino)phenothiazin-5-ium chloride hydrate: 1.0%; Water: 99%
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ06K500 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog Number042771.AP,042771.AE
Lot NumberZ06K501 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog Number042771.AP,042771.AE
Lot NumberX30K500 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number042771.AP,042771.AE
Lot NumberX30K501 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number042771.AP,042771.AE
Lot NumberW26K512 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number042771.AP,042771.AE
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Methylene Blue, 1% w/v aq. soln. > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only