Learn More
IBA Lifesciences MHC I-Strep HLA-A*0201; NY-ESO-1 (SLLMWITQV)
MHC class I molecule fused to a Twin-Strep-tag™ and refolded with a peptide
Brand: IBA Lifesciences 6-7013-001
Description
Specifications
40 μL | |
Fluorescent reagent for staining, magnetic T cell labeling and isolation |