Learn More
Thermo Scientific™ Aluminum Support, tube/plate rack, 384-well
Thermo Scientific™ aluminum support rack can be used for sample preparation, heat-sealing and automatic dispensing.
Brand:  Thermo Scientific™ PRK1383
134.78 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Product Code. 11508525
  • 293.00 € / 1 unit
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Perform sample preparation, heat-sealing and automatic dispensing using Thermo Scientific™ aluminum support rack.
Available in 96- and 384-well format.
  • For heat-sealing and automated dispensing
  • Industry-standard footprint fits in robotic workstations
  • Can be heated or cooled to assist with sample preparation
Includes:
Recommended for:
Sample handling, sample preparation, heat-sealing, automated dispensing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microplate Rack
Microplates (384-well)
Rectangular
1 rack
Blue/Clear, Clear
Aluminum
Real-Time PCR Systems or Thermal Cyclers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Aluminum Support, tube/plate rack, 384-well > For 384-well plates