missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Mild Steel Retort Rods

Structurally strong and resistant to clamp compression scars, these mild steel retort rods are ideal in laboratory and other environments.

Brand:  Nickel Electro™ 8634

Product Code. 12845906

  • 12.90 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Structurally strong and resistant to clamp compression scars
  • Immune to corrosion and rust 
  • Mild steel rod 500mm long for retort stands
  • Retort rod, 10mm x 500mm
Specifications

Specifications

Mild Steel
10 mm
10 mm
500 mm
Retort Rod
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.