Learn More
Fisherbrand™ Mini Electrophoresis System
Pour gel, load contrast strips, and press start. Fisherbrand™ mini Electrophoresis System is ideal for personal use, small laboratories or the classroom and includes gel take, casting sets and power supply.
Brand:  Fisherbrand™ E2102-F-E
Product Code. 15530340
  • 667.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Includes gel box power supply and two casting sets
  • Quick and easy gel casting
  • No clamps or spacers required
  • Compact design
Specifications
Specifications
0 to 99 min. or continuous
Safety lid interlock, two casting sets
190 x 130 x 55 mm
50/60 Hz
One: 105 x 60 mm, Two: 50 x 60 mm
230 mL
115/230 V 50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Mini Electrophoresis System >