Learn More
Endecotts™ Minor 200 Sieve Shaker
Compact and portable. Endecotts Minor 200 Sieve Shaker is ideal for use in laboratories and plants.
Brand: Endecotts™ MIN200/23050
Description
- Compact and portable and easy to use
- Electromagnetic drive for quiet and virtually maintenance free operation
- Requires only small storage space due to small footprint and easily removable clamping belt system (included)
- Different voltages available
Compliance
AASHTO T 27
Specifications
230 V, 50 Hz | |
100/200 mm | |
8 full height/16 half height (200 mm sieves) | |
Analog, 0 to 60 min., Continuous |
Sieve Shaker | |
Minor 200 Benchtop | |
38 μm to 125 mm |