Learn More
Corning™ Modified Impeller Assembly
Stainless steel impeller shaft with modified impeller blade for use with probes to create a small bioreactor.
Brand:  Corning™ 401661
 View more versions of this product
Product Code. 10727993
  • 1156.59 € / Each
Estimated Shipment: 11-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Stainless steel impeller shaft with modified impeller blade for use with probes to create a small bioreactor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Impeller shaft with dual bearing
Stainless Steel
Magnetically-Driven 15L Proculture Spinner Flask
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Modified Impeller Assembly > For use with Magnetically-Driven 15L Proculture Spinner Flask