Learn More
DuPont™ Tyvek 600 Plus White Hooded Coverall
Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ Taped Seam Hooded Coveralls are composed of flash spun high density polyethylene.
Brand:  DuPont™ D13395284
878.23 EUR valid until 2025-12-31
Use promo code "24380" to get your promotional price.
 View more versions of this product
Product Code. 11723083
  • 1045.00 € / Pack of 100
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Self-adhesive chin strap for tight seal of suit to face mask
  • Stitched and over-taped seams for protection and strength
  • Tyvek zipper and zipper flap for enhanced protection
  • Elasticated face, wrists and ankles as well as glued-in elastic waist
  • Elasticated thumb loops prevent sleeves from riding up
  • Permeable to both air and water vapour, yet repels water-based liquids and aerosols
  • Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
  • Applications include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or research, biosecurity laboratories as well as medical applications and when exposed to biological hazards
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hooded Coveralls
HDPE
Elastic
Thumb Loop
Elastic
Stitched, Taped
Chemicals, Liquid
Maintenance and Dismantling Jobs in the Nuclear Industry, Pharmaceutical Manufacturing or in Research and Biosecurity Laboratories
X-Large
White
Elastic
Hooded
Zipper with Self-adhesive Flap and Chin Flap
Category III, Type 4-B, 5-B and 6-B
Category III, Type 4-B, 5-B and 6-B, EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), (EN 1149-5)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
DuPont™ Tyvek 600 Plus White Hooded Coverall > Size: X-Large