Learn More
Invitrogen™ Nile Red
For use with fluorescence microscopy
Brand: Invitrogen™ N1142
225.72 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Nile red is used to localize and quantitate lipids, particularly neutral lipid droplets within cells. Nile red is almost nonfluorescent in water and other polar solvents but undergoes fluorescence enhancement and large absorption and emission blue shifts in nonpolar environments (Excitation/emission maxima ∼552/636 nm in methanol).We recommend dissolving in high-quality anhydrous DMSO or DMF to attain a stock concentration up to 1 mM.
Order Info
Shipping Condition: Room Temperature

Specifications
25 mg | |
Fluorescence Microscope | |
Fluorescence | |
Solid | |
Nile Red Stain |
Room Temperature | |
Cell Membranes & Lipids, Lysosomes | |
C20H18N2O2 | |
Red | |
552/636 nm |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.