Learn More
Invitrogen™ Mouse Beta Endorphin ELISA Kit
ELISA
Brand: Invitrogen™ EEL087
Description
Beta endorphin is an opioid neuropeptide that is produced by neurons of the pituitary gland and released into the circulatory system, or produced by the hypothalamus, and released into the brain and spinal cord through nerve fibers. The precursor polypeptide for beta endorphin is pro-opiomelanocortin.
Specifications
P01193 | |
9.38 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
POMC | |
<10 | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 hrs 20 mins |
15.63 to 1000 pg/mL | |
HRP | |
Plasma, Serum | |
18976 | |
<10% | |
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer | |
96 Tests | |
Plasma 50 μL, Serum 50 μL | |
Beta Endorphin | |
3 hrs 30 mins |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.