Learn More
MP Biomedicals™ γ-Globulin, Human Plasma
Each donor unit used in the preparation of the product was tested and found negative for Anti-HIV 1/2, HBsAg, Anti-HCV and HIV-1 Ag by FDA approved methods or CE approved methods.
Brand:  MP Biomedicals™ 08823102
Product Code. 11414276
  • 395.00 € / 10g
Explore more special offers
Specifications
White
10 g
Powder
>96%
6.44
2°C to 8°C
Used as an immunological challenge in studies of mechanisms of immunotolerance
Human Plasma gamma Globulin
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ γ-Globulin, Human Plasma > Quantity: 10g