missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes w/Sealing Cap
GREENER_CHOICE

View contents easily using these strong, transparent polycarbonate centrifuge tubes, available with sealing caps for use up to 50,000 x g.

Brand:  Thermo Scientific™ 3138-0050

194.96 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 10088810

  • 264.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.

This item is not returnable. View return policy

Description

Description

Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.

Features:
  • Withstand temperatures from -135° to +135°C (-211° to +275°F)
  • Round bottom to fit high speed rotors
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
  • Come with PP sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
  • Autoclavable; transparent

Ordering Information:

Sealing caps also sold separately for 30 and 50mL sizes.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene, red rubber
Polycarbonate
10
Autoclavable
114.8 mm
Screw Cap
28.8 mm
Centrifuge Tube
50 mL
50
Non-sterile
Round
Round
No
No
50,000 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1405285 Certificate TypeCertificate of Compliance Date04/07/2024 Catalog Number3138-0050
Lot Number1399217 Certificate TypeCertificate of Compliance Date25/01/2024 Catalog Number3138-0050
Lot Number1398386 Certificate TypeCertificate of Compliance Date20/12/2023 Catalog Number3138-0050
Lot Number1386590 Certificate TypeCertificate of Compliance Date17/06/2023 Catalog Number3138-0050
Lot Number1378897 Certificate TypeCertificate of Compliance Date01/02/2023 Catalog Number3138-0050
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes w/Sealing Cap > With sealing caps; Capacity: 50mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.