Learn More
Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination with these PETG serum vials. Lighter in weight than glass, they are made for use with lined continuous-thread closures (available separately).
Brand: Thermo Scientific™ 342032-0020
436.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene PETG Serum Vials. These serum vials are lighter in weight than glass, and are made for use with lined continuous-thread closures (available separately).
Features:
- PETG
- Noncytotoxic
- Nonpyrogenic
- Packaged in trayless shrink-wrap modules
- USP VI-compliant
- Sterile to 10-6 SAL
- Leakproof† with Nalgene lined continuous-thread closures (Cat. No. 342158)
Ordering Information:
Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).

Specifications
Serum Vials | |
Clear | |
PETG | |
20 mL | |
153 | |
Nalgene | |
Lined Continuous-thread Closure (Sold Separately) | |
612 Cases, 153 Pk. | |
Serum Vial | |
Lined Continuous-Thread Closure |
11 mm | |
30 mm | |
Sterile | |
612 | |
2/3 oz. | |
20 mL | |
64 mm | |
ISO 13485:2016 | |
20-415 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1420765 | Certificate of Processing | 09/04/2025 | 342032-0020 |
1396948 | Certificate of Processing | 04/12/2024 | 342032-0020 |
1420765 | Certificate of Compliance | 17/01/2025 | 342032-0020 |
1336014 | Certificate of Compliance | 10/12/2024 | 342032-0020 |
1282091 | Certificate of Compliance | 10/12/2024 | 342032-0020 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.