missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
GREENER_CHOICE

Combine these flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.

Brand:  Thermo Scientific™ 4102-0250

70.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10444881

  • 72.80 € / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.

  • Translucent wall makes liquid level easily visible
  • 2000mL size has molded-in graduations
  • Mouth sized to accept standard sized stoppers (stopper sold separately)
  • Repeatedly autoclavable for long life and rigorous use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

8 oz.
250 mL
12
Autoclavable
No
Non-sterile
Stopper No. 6
Flask Erlenmeyer PP 250mL
PPCO
6
Reusable
Narrow
Plain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1419032 Certificate TypeCertificate of Compliance Date08/01/2025 Catalog Number4102-0250
Lot Number1403941 Certificate TypeCertificate of Compliance Date17/04/2024 Catalog Number4102-0250
Lot Number1391982 Certificate TypeCertificate of Compliance Date05/09/2023 Catalog Number4102-0250
Lot Number1360750 Certificate TypeCertificate of Compliance Date29/09/2022 Catalog Number4102-0250
Lot Number1333742 Certificate TypeCertificate of Compliance Date12/10/2021 Catalog Number4102-0250
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks > 250mL, #6 stopper

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.