missing translation for 'onlineSavingsMsg'
Learn More

Nalgene™ Settlometer Jar with Cover
GREENER_CHOICE

Replacement part for the Nalgene™ Settlometer Kit

Brand:  Thermo Scientific Nalgene 711010-9507

203.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10264351

  • 213.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Nalgene™ Settlometer Jar with Cover is a replacement part in the Nalgene™ Settlometer Kit Cat# 1010-0507. The Nalgene™ Settlometer Kit is used to analyze the activated sludge process in waste water treatment plants. The 2L jar is lightweight, transparent, highly impact-resistant and has easy to read white graduations.

Features:

  • Lightweight
  • Transparent
  • Impact-resistant
  • Easy-to-read white graduations
  • Approximate jar dimensions: O.D. × H: 3.3 × 7.5 in. (13.2 × 19.1cm)
  • Polycarbonatematerial
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2L Polycarbonate Jar
Replacement Part
Nalgene™ Settlometer Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nalgene™ Settlometer Jar with Cover >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.