Learn More
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets
GREENER_CHOICE
Offers the highest possible pH precision. Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets are specified to three decimal places and come with a printed certificate that guarantees traceability. Individual packets guarantee fresh solution for every calibration, as well as maximum ease of use.
Brand:  Mettler Toledo™ 30111136
118.35 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 15392265
  • 131.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. Maximum precision is guaranteed with fully traceable NIST/DIN buffers. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
  • NIST/DIN pH buffers are specified and guaranteed to three decimal places, offering top precision
  • Individual sachets guarantee fresh solution for every calibration, as well as maximum ease of use
  • A test certificate and detailed label with a temperature table comes with every box
  • All SDS and labels contain information according to GHS (Globally Harmonized System)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NIST/DIN Buffer pH 4.006 sachets 30x20mL
pH measurement, electrode calibration
Sachet
30 x 20 mL
pH Electrode
Yes
4.006 (at 25°C)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets > NIST/DIN Buffer pH 4.006 sachets 30x20mL