missing translation for 'onlineSavingsMsg'
Learn More

Norell™ HT Plus MHZ Borosilicate Glass NMR Sample Tubes

For auto-sampling systems and high-throughput NMR. Norell™ HT Plus MHZ Borosilicate Glass NMR Sample Tubes have been developed for quality and precision.

Brand:  Norell 552-7

 View more versions of this product

Product Code. 12862336

  • 82.40 € / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Virtually all bottom and wall variation has been eliminated
  • Improved shimmering times and sample reproducibility
  • All tubes have polished ends making them less susceptible to breaking
  • Colored caps are included
Specifications

Specifications

178 mm
NMR Sample Tube
4.97 mm
Routine NMR
5 Pack
Borosilicate Glass
Colorless
4.2 mm
Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Norell™ HT Plus MHZ Borosilicate Glass NMR Sample Tubes > Length: 178mm; Inner Diameter: 4.2mm; Quantity: 5 Pack

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.