Learn More
SYBR™ Power SYBR™ Green PCR Master Mix
Offers superior sensitivity and reproducibility, detecting as few as two copies of a target gene over a broad range of template concentrations.
Brand: SYBR™ 4368577
130.15 EUR valid until 2025-12-31
Use promo code "24271" to get your promotional price.
Includes: SYBR™ Green 1 Dye, AmpliTaq Gold DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components
Requires : Template and primers
This item is not returnable. View return policy
Description
Get superior nucleic acid quantitation with Applied Biosystems™ Power SYBR™ Green Master Mix.
- Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient single-tube format
- Power SYBR™ Green PCR Master Mix contains all of the components (excluding the template and primers) for superior SYBR™ Green reagent-based real-time PCR
- The optimized formulation contains highly purified AmpliTaq Gold™ DNA Polymerase, LD to offer increased sensitivity
- Contains a blend of dTTP/dUTP that is compatible with AmpErase™ UNG to minimize carryover contamination
- Includes a proprietary version of ROX™ dye, an internal passive reference, to normalize non-PCR–related fluorescence fluctuations
- This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error and sample evaporation
- Power SYBR™ Green reagent-based PCR chemistry easily replaces the existing SYBR™ Green PCR Master Mix in Applied Biosystems™ protocols using the same setup and thermal cycling conditions
- Store at -20°C
Order Info
Shipping Condition: Wet Ice

Specifications
The 2x mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ DNA Polymerase LD, dNTPs with dUTP⁄dTTP blend, Passive Reference 1 and optimized buffer components. Contains 1 X 1 mL vial, sufficient for 40 reactions at 50 μL. Store at -20°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Tube | |
qPCR | |
Standard | |
SYBR Green | |
SYBR™ | |
LD (Low DNA) | |
Wet Ice | |
Gene Expression | |
40 Reactions | |
1 x 1 mL |
SYBR | |
High | |
AmpliTaq Gold DNA Polymerase, AmpliTaq Gold LD | |
7300 System, 7500 System, 7700 System, 7900HT Fast System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
1 x 1 mL | |
40 Reactions of 50 μL | |
2X | |
DNA (Genomic), cDNA |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2502198 | Certificate of Analysis | 14/03/2025 | |
2411196 | Certificate of Analysis | 27/11/2024 | 4368577,4368577_LITL18 PROMO |
2407195 | Certificate of Analysis | 16/08/2024 | 4368577,4368577_LITL18 PROMO |
2405194 | Certificate of Analysis | 30/05/2024 | 4368577,4368577_LITL18 PROMO |
2403193 | Certificate of Analysis | 18/03/2024 | 4368577,4368577_LITL18 PROMO |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.