missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Polycarbonate Cell Culture Inserts in Multi-Well Plates
BIOPHARMA

Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.

268.00 € - 500.00 €

Specifications

Material Polycarbonate (Membrane)
Type Culture Insert
Color Clear
Surface Treatment Cell culture-treated
Sterility Sterile
View More Specs

Products 12
Product Code Brand Pore Size Dish Type Culture Area Price Quantity & Availability  
Product Code Brand Pore Size Dish Type Culture Area Price Quantity & Availability  
10387523
BIOPHARMA
View Documents
Thermo Scientific™
140620
0.4 μm 24-Well Plate 0.47 cm2
500.00 €
Pack of 48
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
10021863
BIOPHARMA
View Documents
Thermo Scientific™
140627
3 μm 24-Well Plate 0.47 cm2
448.00 €
Pack of 48
In Stock
to see stock.
Add to basket
 
10002433
BIOPHARMA
View Documents
Thermo Scientific™
140629
8 μm 24-Well Plate 0.47 cm2
500.00 €
Pack of 48
In Stock
to see stock.
Add to basket
 
10543624
BIOPHARMA
View Documents
Thermo Scientific™
140640
0.4 μm 6-Well Plate 3.14 cm2
269.00 €
Pack of 24
In Stock
to see stock.
Add to basket
 
10092153
View Documents
Thermo Scientific™
140642
3 μm 6-Well Plate 3.14 cm2
270.00 €
Pack of 24
In Stock
to see stock.
Add to basket
 
10637143
View Documents
Thermo Scientific™
140644
8 μm 6-Well Plate 3.14 cm2
269.00 €
Pack of 24
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
 
10478803
View Documents
Thermo Scientific™
140652
0.4 μm 12-Well Plate 1.13 cm2
460.00 €
Pack of 48
Estimated Shipment: 23-12-2024
to see stock.
Add to basket
 
10504394
BIOPHARMA
View Documents
Thermo Scientific™
140654
3 μm 12-Well Plate 1.13 cm2
460.00 €
Pack of 48
In Stock
to see stock.
Add to basket
 
10236103
View Documents
Thermo Scientific™
140656
8 μm 12-Well Plate 1.13 cm2
459.00 €
Pack of 48
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
 
10124093
View Documents
Thermo Scientific™
140660
0.4 μm 6-Well Plate 4.1 cm2
269.00 €
Pack of 24
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
 
10276803
View Documents
Thermo Scientific™
140663
3 μm 6-Well Plate 4.1 cm2
270.00 €
Pack of 24
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
 
10377143
View Documents
Thermo Scientific™
140668
8 μm 6-Well Plate 4.1 cm2
268.00 €
Pack of 24
In Stock
to see stock.
Add to basket
 
Description

Description

Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.

Features:
  • Tissue culture-treated porous membrane for optimized cell attachment
  • Three pore sizes: 0.4, 3, and 8 micron
  • Pre-packed in sterile Nunclon™ Delta-treated multi-well plates
  • Nontoxic and nonpyrogenic

Applications:

  • Tumor migration and invasion assays
  • Epithelial (e.g. Caco-2) and endothelial (e.g. brain microvascular endothelial cells) barriers assays
  • Dermal/epidermal and epithelial tissue models (e.g. artificial skin model)
  • Chemotaxis studies
  • Co-culture

Warranty: 90 days

 

Specifications

Specifications

Polycarbonate (Membrane)
Clear
Sterile
Culture Insert
Cell culture-treated
Videos
SDS
Documents

Documents

Product Certifications

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.