Learn More
Macherey-Nagel™ Optima™ FFAP GC Capillary Column
Brand:  Macherey-Nagel™ 726116.25
 View more versions of this product
Product Code. 10398251
  • 605.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
0.25 mm
G1, G2, G38
240°C to 250°C
25 m
GC Capillary Column
Optima™ FFAP
Free Carboxylic Acids Analysis, Free Carboxylic Acids
0.25 μm
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Optima™ FFAP GC Capillary Column > Length: 25m; I.D.: 0.25mm; Film Thickness: 0.25um