Learn More
Novacyt - Microgen Bioproducts PathFlow Adenovirus Test Kit
The Microgen Bioproducts PathFlow Adenovirus antigen is a rapid chromatographic immunoassay for the qualitative detection of adenovirus.
Brand: Novacyt - Microgen Bioproducts M592CE
Description
The Cassette is a qualitative membrane-based immunoassay for the detection of adenovirus antigen in eye conjunctive swabs, throat swabs and nasal swabs.
- Simple and easy to use system
- Differential detection of adenovirus from other diseases preventing similar symptoms
- Rapid results offered; positive results may be visible from as little as 3 minutes
- Can be used with multiple sample types, including eye conjunctive swabs and throat/nasopharyngeal swabs
Specifications
ISO13485 ISO9001 | |
Yes | |
Anti-Adenovirus antibody particles and anti-Adenovirus antibodies | |
Swab | |
Test Strip | |
Timer | |
Plastic | |
Animal | |
20 Tests and 1 Bottle (10 mL) | |
Adenovirus | |
Eukaryota |
2°C to 30°C | |
Non-waived | |
20 Tests | |
0.986 | |
The Qualitative Detection of Adenovirusin Eye Conjunctive Swab, Throat Swab and Nasal Swabspecimen | |
20 Tests | |
1/Test | |
Carton with sealed foil pouches | |
Contains anti-Adenovirusantibody particles and anti-Adenovirusantibodiescoated on the membrane | |
0.981 | |
15 minutes |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.