Learn More
Thermo Scientific™ Amino Acid Standard H
Calibrate HPLC analysis of protein hydrolysates using this calibration standard mixture of 18 amino acids, each at 2.5 micromoles/mL in 0.1N HCl.
Brand: Thermo Scientific™ 20088
171.00 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ Pierce Amino Acid Standard H is a quantitative mixture of 18 amino acids, supplied at 2.5μmol/mL in 0.1N HCl, for use as a high-purity calibration standard for HPLC analysis of protein hydrolysates.
Highlights:
- Uses L-form configuration to permit standardization of microbial and other assays
- Molar concentration verified by conventional amino acid analysis methods
- With the exception of cystine, each amino acid is supplied at a concentration of 2.5μmoles/mL in 0.1N HCl
- L-alanine, Ammonia [(NH4)2SO4], L-Arginine, L-Aspartic Acid, L-Cystine, L-Glutamic Acid, Glycine, L-Histidine, L-Isoleucine, L-Leucine, L-Lysine·HCl, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tyrosine, L-Valine
Includes:
- L-alanine, Ammonia [(NH4)2SO4], L-Arginine, L-Aspartic Acid, L-Cystine, L-Glutamic Acid, Glycine, L-Histidine, L-Isoleucine, L-Leucine, L-Lysine·HCl, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tyrosine, L-Valine

Specifications
Amino Acid Standard H | |
Store between -20°C and 8°C. | |
10 x 1 mL | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZJ399988 | Certificate of Analysis | 16/12/2024 | 20088,TS-20088 |
ZG393876 | Certificate of Analysis | 07/08/2024 | 20088,TS-20088 |
ZE388332 | Certificate of Analysis | 11/06/2024 | 20088,TS-20088 |
AA381507 | Certificate of Analysis | 25/01/2024 | 20088,TS-20088 |
SPECS | Certificate of Analysis | 25/01/2024 | 20088,TS-20088 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.