missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ PCR 8-Strip Caps

Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples.PCR 8-Strip Caps are autoclavable and RNase-, DNase-, and DNA-free. Domed caps are available in a variety of colors for sample identification.

Brand:  BRAND™ 781334

40.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10271783

  • 46.30 € / Pack of 125

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.
  • Available flat or domed
  • The domed caps are available in a variety of colors for sample identification
  • Autoclavable
  • RNase-, DNase-, and DNA-free.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Non-sterile
Yes
8-strip Flat Cap
Yes
Flat
Microplates
Polypropylene
Yes
Autoclavable
Single-use
Cap Strip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Brochures

Technical Notes

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number0000012929 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number781334
Lot Number0000012605 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number781334
Lot Number0000010594 Certificate TypeCertificate of Analysis Date21/05/2024 Catalog Number781334
Lot Number0000010399 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number781334
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ PCR 8-Strip Caps > PCR 8-strip flat caps; Clear

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.