Learn More
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Brand: Macherey-Nagel™ 723341.25
Specifications
0.5 mm | |
0.32 mm | |
G25, G35 | |
220°C to 240°C | |
25 m | |
1 Each |
GC Capillary Column | |
Permabond™ FFAP | |
Free Carboxylic Acids Analysis, Free Carboxylic Acids | |
0.25 μm | |
Permabond™ FFAP |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.