Learn More
Macherey-Nagel™ pH Indicator Papers, Refill
The longstanding, appreciated standard for many pH testing applications.
Brand: Macherey-Nagel™ 90233
This item is not returnable.
View return policy
Description
- pH indicator papers have been on the market for decades and are the appreciated standard for many applications
- For each pH value these papers show a single color which can be matched with the color scale at intervals of 0.2–1 pH unit
Specifications
9 to 13 | |
pH Indicator Paper, Refill | |
7 mm |
1 x 5 m x 7 mm Spool | |
5 m |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Macherey-Nagel™ pH Indicator Papers, Refill > pH 9.0 to 13.0
Spot an opportunity for improvement?Share a Content Correction