Learn More
Cytiva PhastGel™ Precast IEF Gels
Precast homogeneous gels containing Pharmalyte as carrier ampholyte to generate stable, linear pH gradients with uniform conductivity across the entire pH range and allow high field strengths for high resolution separations.
Brand:  Cytiva 17-0543-01
 View more versions of this product
Product Code. 10678225
Description
Description
  • PhastGel IEF gels
  • Homogeneous gels in various pH ranges for separations of proteins by isoelectric point
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva PhastGel™ Precast IEF Gels > IEF 3-8