Learn More
DWK Life Sciences Wheaton™ Crimping Jaw Set for High Performance Crimping Tool
Interchangeable jaws allow users to choose a jaw that best fits their needs.
Brand: DWK Life Sciences W225771
775.17 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps
Specifications
High performance crimping tool | |
20 mm | |
Crimping |
Product Suggestions
Customers who viewed this item also viewed.