Learn More
Cytiva PhastGel™ Precast IEF Gels
Precast homogeneous gels containing Pharmalyte as carrier ampholyte to generate stable, linear pH gradients with uniform conductivity across the entire pH range and allow high field strengths for high resolution separations.
Brand: Cytiva 17-0545-01
View more versions of this product
Description
- PhastGel IEF gels
- Homogeneous gels in various pH ranges for separations of proteins by isoelectric point

Specifications
125°C | |
PhastGel IEF – 5-8 | |
0.45 mm | |
pH 5 to 8 | |
Protein | |
Isoelectric Focusing | |
Charecterizing and Analysing Mixtures of Proteins with Smaller PI Differentials | |
82° to 86°C | |
10/Pk. | |
0.017 in. | |
4.8 cm | |
1.9 in. | |
5 cm | |
4.8 cm |
Polyester | |
2 to 6% | |
Pharmalyte | |
IEF Gel | |
pH 5 to 8 | |
Containing 2 to 6% Pharmalyte as carrier Ampholyte | |
GelBond polyester film | |
71.08 | |
4 - 8°C | |
50 mm | |
1.9 in. | |
48 mm | |
1.88 in. |
Safety and Handling
- Danger
Hazard Category
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
Hazard Statement
- H340-May cause genetic defects.
- H350-May cause cancer.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Acrylamide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.