Learn More
Phloroglucinol, anhydrous, 98%, Thermo Scientific Chemicals
Description
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .
Solubility
Soluble in diethyl ether, ethanol and pyridine.
Notes
Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.

Specifications
Specifications
Melting Point | 215°C to 220°C |
Quantity | 50 g |
Beilstein | 1341907 |
Sensitivity | Hygroscopic; Light sensitive |
Merck Index | 14,7328 |
Solubility Information | Soluble in diethyl ether,ethanol and pyridine. |
Formula Weight | 126.11 |
Percent Purity | 98% |
Chemical Name or Material | Phloroglucinol, anhydrous |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z07J025 | Certificate of Analysis | 30/07/2024 | B25502.18,B25502.30 |
M31K031 | Certificate of Analysis | 08/02/2024 | B25502.18,B25502.30 |
10244426 | Certificate of Analysis | 03/11/2022 | B25502.18,B25502.30 |
5011A14W | Certificate of Analysis | 23/11/2021 | B25502.18,B25502.30 |
10188454 | Certificate of Analysis | 23/11/2021 | B25502.18,B25502.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.