missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FDAA (Marfey's Reagent) (1-fluoro-2-4-dinitrophenyl-5-L-alanine amide)

Label and measure optical isomers of amino acids by reverse-phase chromatography (HPLC) with this amine-reactive, colored (Amax 340nm) compound.

Brand:  Thermo Scientific™ 48895

148.20 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.



Product Code. 11889380

  • 158.00 € / 50mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce FDAA (Marfey's Reagents) reacts with primary amines to enable the separation and quantitation of optical isomers of amino acids by reverse-phase chromatography.

Highlights:

  • Optical isomers of amino acids derivatization complete in 90 minutes
  • Derivatives have an absorption coefficient of ∽3 x 104 Derivatives can be detected by UV at 340nm with picomole sensitivity
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FDAA
Upon receipt store product at room temperature.
50 mg
Amine
Other Label or Dye
Solid
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA401355 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number48895,TS-48895
Lot NumberZI399026 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number48895,TS-48895
Lot NumberZH395725 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number48895,TS-48895
Lot NumberZD393444 Certificate TypeCertificate of Analysis Date11/05/2024 Catalog Number48895,TS-48895
Lot NumberZB391270 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number48895,TS-48895
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ FDAA (Marfey's Reagent) (1-fluoro-2-4-dinitrophenyl-5-L-alanine amide) > FDAA, Marfey' s Reagent; 50mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.