missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ S-Nitrosylation Western Blot Kit and HENS Buffer

Thermo Scientific Pierce S-Nitrosylation Western Blot Kit enables sensitive detection of protein S-nitrosocysteine post-translational modifications in a complete, easy-to-use kit or stand-alone HENS buffer for S-Nitrosylation Western Blot Kit.

Brand:  Thermo Scientific™ 90105

 View more versions of this product

Product Code. 13464199

  • 552.00 € / 0.20mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Pierce™ S-Nitrosylation Western Blot Kit
Pierce™
All species
S-Nitrosylation
40 Reactions
Upon receipt store kit at –20°C, or store individual components as indicated above. Kit is shipped with an ice pack.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL393742 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number90105
Lot NumberZC386480 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number90105
Lot NumberYJ369011 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog Number90105
Lot NumberYD359197 Certificate TypeCertificate of Analysis Date26/04/2023 Catalog Number90105
Lot NumberYB359196 Certificate TypeCertificate of Analysis Date03/03/2023 Catalog Number90105
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ S-Nitrosylation Western Blot Kit and HENS Buffer > Pierce™ S-Nitrosylation Western Blot Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.