missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Plug-In Tube Extension for UF30, Straight

Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.

Brand:  Memmert™ UF30B29719

Product Code. 17599215

  • 129.00 € / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications

Specifications

UF30 Oven
Plug-in Tube Extension
O.D. 60.3 mm, I.D. 57 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Memmert™ Plug-In Tube Extension for UF30, Straight > For Use With: UF30

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.