Learn More
BRAND™ Polypropylene Photographic Trays
White stackable polypropylene (PP) photographic trays with ribbed bottom that improves stability.
13.10 € - 118.00 €
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
10643782
|
BRAND™
156640 |
45 mm |
13.10 €
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
10706711
|
BRAND™
156650 |
160 mm |
118.00 €
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
10499712
|
BRAND™
156644 |
75 mm |
33.70 €
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
10558591
|
BRAND™
156642 |
70 mm |
23.65 €
Each |
In Stock
Log in to see stock. |
|||||
10409722
|
BRAND™
156646 |
120 mm |
46.75 €
Each |
In Stock
Log in to see stock. |
|||||
10079850
|
BRAND™
156648 |
140 mm |
80.95 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- White stackable polypropylene (PP) photographic trays
- Ribbed bottom improves stability, reinforced rim, slightly sloping walls
- Autoclavable (121 °C)