Learn More
Invitrogen™ Power Blotter XL System
Get rapid and efficient semi-dry protein transfer with the Invitrogen™ Power Blotter.
Brand:  Invitrogen™ PB0013
Product Code. 15816162
  • 3750.00 € / Each
Explore more special offers
Includes: Invitrogen Power Blotter XL Cassette, Invitrogen Power Blotter Station
Description
Description
The Invitrogen™ Power Blotter System is specifically designed for rapid semi-dry transfer of proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter includes an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
The Power Blotter Cassette enables the simultaneous transfer of gels. The Power Blotter Station has an easy-to-use color LCD touchscreen interface with pre-programmed transfer methods for different numbers and sizes of gels and ranges of molecular weight proteins. The easy-touch programming feature allows custom transfer settings to be quickly created, saved, and run.
The Power Blotter achieves highly efficient transfer in 7 minutes when used with Power Blotter Select Transfer Stacks or 10 minutes with the Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional Towbin buffer.
Features:
  • Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
  • Fast—5 to 12 minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
  • Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
  • Easy-touch programming—convenient pre-programmed transfer methods or create, save, and run customized transfer methods
  • Flexible—simultaneous transfer
  • Versatile—compatible with Power Blotter Select Transfer Stacks, Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• Power Blotter Station
• Power Blotter Cassette XL
Nitrocellulose, PVDF
Power Blotter
Blotter System
Room Temperature
Western Blot
Semi-Dry
2 Midi or 4 Mini Gels
1 system
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Power Blotter XL System > Includes: Power Blotter XL Cassette; Power Power Station