Learn More
Applied Biosystems™ PowerUp™ SYBR™ Green Master Mix for qPCR
Pre-formulated, optimized, universal 2X master mix for real-time PCR workflows.
Brand: Applied Biosystems™ A25777
1529.50 EUR valid until 2025-12-31
Use promo code "24271" to get your promotional price.
Description
Coupled with user-supplied primer sets and template, PowerUp™ SYBR™ Green Master Mix is designed to amplify targets for accurate gene expression analysis.
- A dual hot-start mechanism for excellent specificity
- Highly reproducible CTs over a broad dynamic range
- Inclusion of UDG to help prevent carryover contamination
- Stability of pre-assembled reactions for up to 72 hours
- Compatibility with most real-time qPCR instruments
Exceptional specificity with dual hot-start mechanism:
- Specificity is paramount in obtaining high-quality data in SYBR™ Green reactions and can be enhanced through control of the Taq DNA polymerase at lower temperatures before stringent primer binding
- PowerUp™ SYBR™ Green Master Mix uses the Dual-Lock™ Taq DNA polymerase enzyme that combines two unique hot-start mechanisms to help control its activity and to prevent undesirable early activity of the polymerase at low temperatures
- In an experiment evaluating 24 different primer sets, PowerUp™ SYBR™ Green Master Mix generated single melt curves 100% of the time, consistent with highly specific amplification
Tight reproducibility in CTs over a wide dynamic range:
- PowerUp™ SYBR™ Green Master Mix demonstrates excellent reproducibility over a wide dynamic range for a variety of targets tested
- In addition, this master mix provides efficient amplification over 6 logs of sample input
- Unlike competitor mixes that can exhibit inhibition at high cDNA inputs and low correlation coefficients over a dynamic range experiment, PowerUp™ SYBR™ Green Master Mix efficiency is maintained over the full dynamic range to help deliver maximal confidence in data quality
UDG included for carryover contamination control:
- The inclusion of uracil-DNA glycosylase (UDG) and dUTP in the PowerUp™ SYBR™ Green Master Mix enables the degradation of any previously amplified PCR products, helping prevent contamination of subsequent qPCR reactions and possible false positives
Stability of pre-assembled reactions:
- The tight control of the Dual-Lock™ Taq DNA polymerase in PowerUp™ SYBR™ Green Master Mix enables reactions to be assembled up to 72 hours prior to cycling without impacting data
- This consistency allows users to have confidence and flexibility in the use of PowerUp™ SYBR™ Green Master Mix across numerous workflow scenarios1
Broad instrument compatibility:
- PowerUp™ SYBR™ Green Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems™ real-time PCR instruments2
- It is also compatible with the Bio-Rad IQ™5 and CFX96™/CFX384™ systems, Roche LightCycler™ LC480, and Stratagene™ MX3005P™ systems
- For optimal results, recommended primer concentrations are 300 to 800nM
Order Info
Shipping Conditions: Wet Ice

Specifications
The 2X mix contains SYBR Green dye, Dual-Lock Taq DNA Polymerase, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye, and optimized buffer components. Contains 5 x 5 mL tube, sufficient for 2500 20-μL reactions (10 μL Master Mix per reaction). Store at 2–8°C in the dark. |
|
Tube | |
Dual-Lock Taq DNA Polymerase | |
7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 7 Flex, StepOne™, StepOnePlus™, ViiA™ 7 System | |
Real Time PCR SYBR Master Mix | |
Wet Ice | |
Gene Expression | |
2500 Reactions | |
5 x 5 mL |
SYBR | |
qPCR | |
Fast or Standard | |
PowerUP™, SYBR™ | |
5 x 5 mL | |
2500 Reactions of 20-μL | |
2X | |
DNA |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.