Learn More
Thermo Scientific™ ProSpecT™ Giardia Controls
Rapidly detect Giardia specific antigen from human fecal samples.
Brand: Thermo Scientific™ R24280
Description
Rapidly detect Giardia specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Giardia Microplate Assays with ProSpecT™ Giardia Controls. Package contains two positive and two negative control vials for use with Giardia assays.
Manage your resources, increase productivity and improve workflow efficiency with a direct, qualitative enzyme immmunoassay for the detection of Giardia. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.- Excellent clinical performance
- Cary Blair transport option which saves a dilution step
- Results within 100 minutes
- Room temperature incubation
- True colorless negatives
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
2°C to 8°C | |
Giardia | |
Giardia Antigen Testing | |
Two positive and two negative control vials (0.75mL) |
ProSpecT Giardia Controls | |
Fecal | |
Enzyme Immunoassay Detection | |
4 Vials/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2263625 | Certificate of Analysis | 05/07/2018 | R24280 |
2108915 | Certificate of Analysis | 22/06/2018 | R24280 |
1273971-COA | Certificate of Analysis | 15/04/2017 | R24280 |
1472746-COA | Certificate of Analysis | 15/04/2017 | R24280 |
1260015-COA | Certificate of Analysis | 15/04/2017 | R24280 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.