missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ PTFE-Lined Closed Caps Product Code.: 16638482
Estimated Shipment: 01-05-2025
to see stock.
Add to basket

Thermo Scientific™ PTFE-Lined Closed Caps
GREENER_CHOICE

Product Code. 16638482
89-400 mm, Unprocessed, 24, Polypropylene
Click to view available options
Closure Size:
58-400 mm
83-400 mm
89-400 mm
Level:
Unprocessed
No. per Case:
24
Material:
Polypropylene
This item is not returnable. View return policy

Product Code. 16638482

Brand: Thermo Scientific™ B1678900BL

Estimated Shipment: 01-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Use these caps as replacements for our glass environmental sample bottles, jars, jugs and vials.

Thermo Scientific™ PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials. The same high quality PTFE-lined caps used on our Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.

  • White polypropylene closed top cap with bonded PTFE liner
  • 24-414 cap has a PTFE-faced silicone liner for added cushion and an extra secure seal when used with bottles and VOA vials

Includes:

Cap with bonded liner

Compatible with:

For use with Thermo Scientific glass environmental sample bottles, jars, jugs and vials.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Certification Parameters None
Color White
Description Closed top cap
Material Polypropylene
No. per Case 24
Closure Color White
Level Unprocessed
Liner Material PTFE
Certifications/Compliance Manufactured under an ISO certified quality management system.
Closure Included Yes
Shape Round
Product Type Closure with Bonded Liner
Closure Liner PTFE
Closure Material Polypropylene
Closure Size 89-400 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ PTFE-Lined Closed Caps >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.