missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PVC Reference Tubing for MF1 and MF4 Pump Heads

For use with MF1 and MF4 Pump Heads

Brand:  Gilson™ F117940

 View more versions of this product

Product Code. 10581093

  • 106.00 € / Pack of 12
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Calibrated
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
PVC
For MF1 and MF4 Pump Heads
12 Pack
Low
94°C
1.29 mm
40 cm
General use
9mL/min.
Transparent
Pump Tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilson™ PVC Reference Tubing for MF1 and MF4 Pump Heads > I.D.: 1.29mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.