Learn More
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper
Manufactured from chemically resistant Pyrex™ borosilicate glass with hexagonal glass.
Brand:  Pyrex 3212/18WC
 View more versions of this product
Product Code. 15767957
  • 270.00 € / Each
Estimated Shipment: 25-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complies with ISO 4788
  • Calibrated to Class A tolerances
  • Individually tested and supplied with certificate showing exact volume contained at 5 points on the scale
  • Conformity H certified
  • Printed with individual serial number for batch certification
  • Manufactured from chemically resistant Pyrex borosilicate glass with hexagonal glass
  • Graduations in white enamel for improved visibility with dark colored solutions
  • Supplied with polyethylene stopper
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hexagonal
Glass
A
545 mm
45/40
Graduated Cylinder
ISO 4788
2000 mL
Polyethylene Stopper
Polyethylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper > Capacity: 2000mL