Learn More
Pyrex™ Borosilicate Glass Cylinders in Squat Format
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Squat Form are ideal for use in fume hoods and restricted spaces. Borosilicate glass gives superior chemical resistance.
Brand:  Pyrex 3285-1L
 View more versions of this product
Product Code. 12632042
  • 258.00 € / Pack of 2
Estimated Shipment: 08-10-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hexagonal base and squat form delivers stability
  • Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
  • Heavily beaded rim and spout allow clean and easy dispensing
  • Class B
  • Complies with ISO 4788
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hexagonal
Round
±10 mL
BS 604, DIN 12680, ISO 4788
1000 mL
289 mm
20 mL
Blue
Graduated Cylinder
Borosilicate Glass
B
Confined Spaces
Blue enamel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pyrex™ Borosilicate Glass Cylinders in Squat Format > Capacity: 1000mL; Graduations: 20mL