Learn More
Applied Biosystems™ QuantStudio™ 12K Flex Real-Time PCR Software
QuantStudio™ 12K Flex Software supports the QuantStudio™ 12K Flex Real-Time PCR System
Brand:  Applied Biosystems™ 4472048
Product Code. 15779714
  • 333.00 € / Each
Explore more special offers
Description
Description
  • QuantStudio™ 12K Flex Software supports the QuantStudio™ 12K Flex Real-Time PCR System.
  • The software allows the user to open and analyze experiments generated from the QuantStudio™ 12K Flex Real-Time PCR System.
  • The software also enables the user to set up experiments, send experiments to the instrument, control the thermal cycling process in the instrument, collect data and analyze the collected data, all in an integrated and streamlined fashion.
  • This configuration includes a license kit with 10 keys for 10 users. Download the software installer (a license key is required for use)
  • QuantStudio™ System home screen enables launch of analysis applications from within application
  • New Sample Tracker software for OpenArray™ plates
  • Easy to set up experiments for both novice and advanced users for 384-well and OpenArray™ blocks
  • Customizable optical filter selection for more complex applications, such as multiplexing
  • QuickStart setup so you can begin a run immediately and enter plate information later
  • Ability to run up to four OpenArray™ plate experiments at once
  • Real-time monitoring of amplification curves enables you to view a run progress (can be viewed from a remote PC as well)
  • Auto-baseline and auto-threshold for simplified data analysis
  • Alternative method for data analysis using a non-threshold CT calling method
  • Multiple Plots view for simultaneously assessing data from four perspectives
  • Automated SNP genotype calling with intuitive graphical output and quality-value assignment
  • Troubleshooting flags to help you diagnose and solve problematic experiments
  • Tool tips for easy identification of sample wells when viewing amplification curves or SNP genotyping plots
  • Email notifications to alert you when a run has started or ended
  • Easy cut-and-paste functionality
  • export easily to Microsoft™ PowerPoint™ software, Excel™ software, or directly as a .JPEG file
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Real-Time PCR Data Analysis
QuantStudio 12k Flex
Windows 7 32-Bit
10 seats
PCR Management, QuantStudio™ 12K Flex RTPCR System
10 Seat
QuantStudio
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ QuantStudio™ 12K Flex Real-Time PCR Software > 10 seats