Learn More
Applied Biosystems™ QuantStudio™ Absolute Q™ MAP16 Plate Kit and Master Mix
A key component of the QuantStudio Absolute Q Digital PCR System powered by the patented microfluidic array plate (MAP) technology, enabling consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV).
Brand:  Applied Biosystems™ A53301
705.16 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Product Code. 17427823
  • 1178.00 € / Each
Edge
Explore more special offers
Description
Description
  • A key component of the QuantStudio Absolute Q Digital PCR System
  • System is powered by the patented microfluidic array plate (MAP) technology that enables consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV)
  • QuantStudio Absolute Q MAP16 consumable uses microinjection-molded plates to overcome common challenges of inconsistency and high dead volume
  • Benefits of this approach include precise microchamber volume, consistent sample distribution, and greater than 98% of loaded volume analyzed
  • Includes QuantStudio Absolute Q DNA Master Mix optimized for use with the QuantStudio Absolute Q Digital PCR System and Applied Biosystems digital PCR (dPCR) assays
  • 5X formulation enables analysis of higher sample volumes and delivers accurate quantification of DNA targets without using a standard curve
  • Unparalleled precision and accuracy of the Absolute Q system provides confidence in measurements across replicates and experiments
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Includes Barcode
Digital PCR
12 MAP16 Plates, Gaskets, Isolation Buffer, and Absolute Q™ DNA Digital PCR Master Mix
12 plates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ QuantStudio™ Absolute Q™ MAP16 Plate Kit and Master Mix > Digital PCR
For Research Use Only. Not for use in diagnostic procedures