Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand:  Quickfit XA41
 View more versions of this product
Product Code. 11928044
  • 112.00 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
  • Useful for the connection of dissimilar sized ground glass joints
  • Large sized socket at the top and small sized cone at the base
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14/23
Borosilicate Glass
29/32
Useful for the connection of dissimilar sized ground Glass joints
Expansion adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Expansion Socket Adapter > Cone size: 14/23; Socket size: 29/32