Learn More
R&D Systems™ Malachite Green Phosphate Detection Kit
contains sufficient materials to run Sandwich ELISAs on at least 1 Kit
Brand: R&D Systems™ DY996
250.40 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
Description
- Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
- Development protocols are provided to guide further assay optimization
- Assay can be customized to your specific needs
- Economical alternative to complete kits

Specifications
Malachite Green Phosphate Detection Kit | |
Phosphate Detection | |
1 Kit | |
Store at room temperature |
Fifteen 96-well plates | |
Malachite Green Reagent A (Ammonium molybdate in 3M sulfuric acid); Malachite Green Reagent B (Malachite green oxalate and polyvinyl alcohol); Phosphate Standard | |
50 μL | |
4 hours 40 minutes (after plate preparation) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.