missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapID™ Nitrate A Reagent

Complete the Thermo Scientific™ RapID™ System Nitrite test using the ready-to-use Thermo Scientific RapID Nitrate A Reagent.

Brand:  Thermo Scientific™ R8309003


Product Code. 13285559

  • 33.30 € / 15mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Complete the Thermo Scientific™ RapID™ System Nitrite test using the Thermo Scientific RapID Nitrate A Reagent.

This product is for use with RapID CB PLUS System (R8311008), RapID NF PLUS System (R8311005), RapID ONE System (R8309002), RapID NH System (R8311001), RapID STAPH PLUS System (R8309003) and RapID NH System (R8311001).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 8°C
Yes
RapID CB PLUS System (R8311008), RapID NF PLUS System (R8311005), RapID ONE System (R8309002), RapID NH System (R8311001), RapID STAPH PLUS System (R8309003, RapID NH System (R8311001)
15 mL/Vial
RapID Nitrate A Reagent
Vial
1 Box
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents

Documents

Instructions

Manuals

Specification Sheets

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6212721 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberR8309003
Lot Number6195493 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberR8309003
Lot Number6155300 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberR8309003
Lot Number6128848 Certificate TypeCertificate of Analysis Date10/09/2024 Catalog NumberR8309003
Lot Number3780796 Certificate TypeCertificate of Analysis Date06/04/2024 Catalog NumberR8309003
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ RapID™ Nitrate A Reagent > 15mL/Vial

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.