Learn More
Thermo Scientific Chemicals RBC lysis buffer for mouse
For lysis of mouse red blood cells
Brand: Thermo Scientific Chemicals J62150.AK
61.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
RBC Lysis Buffer has been reported for use in flow cytometric analysis and cell culture. Red Blood Cell Lysing Buffer has been developed for use in hybridoma protocols to remove red blood cells from mouse splenocyte suspensions before fusion. It is also useful in systems where it may be desirable to remove red blood cells from cell suspensions, such as whole blood.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
250mL | |
Cell Lysis Reagent | |
2° to 8°C |
Liquid | |
For lysis of mouse red blood cells |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P15L516 | Certificate of Analysis | 14/03/2025 | J62150.AP,J62150.AK |
P15L517 | Certificate of Analysis | 14/03/2025 | J62150.AP,J62150.AK |
W23K517 | Certificate of Analysis | 30/10/2024 | J62150.AP,J62150.AK |
X01K547 | Certificate of Analysis | 30/10/2024 | J62150.AP,J62150.AK |
R09K513 | Certificate of Analysis | 26/06/2024 | J62150.AP,J62150.AK |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only