Learn More
BINDER™ Reinforced Rack
Reinforced shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
Brand:  BINDER™ 8012-2412
 View more versions of this product
Product Code. 30238242
  • 291.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Shelf rack made of stainless steel
  • Includes set of U-rails
Specifications
Specifications
KBF 130, KBF PRO 130
Stainless Steel
Rack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Reinforced Rack > KBF 130, KBF PRO 130